Learn More
Medchemexpress LLC HY-103414 10mg Medchemexpress, Raclopride CAS:84225-95-6 Purity:>98%
SDP
Catalog No. 501968105
Encompass_Preferred
Medchemexpress, HY-103414 10mg Raclopride CAS:84225-95-6 Raclopride is a dopamine D2/D3 receptor antagonist, which binds to D2 and D3 receptors with dissociation constants (Kis) of 1.8 nM and 3.5 nM, respectively, but has a very low affinity for D1 and D4 receptors with Kis of 18000 nM and 2400 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8105 Supplier Medchemexpress LLC Supplier No. HY10341410MG
Medchemexpress, HY-103414 10mg Raclopride CAS:84225-95-6 Raclopride is a dopamine D2/D3 receptor antagonist, which binds to D2 and D3 receptors with dissociation constants (Kis) of 1.8 nM and 3.5 nM, respectively, but has a very low affinity for D1 and D4 receptors with Kis of 18000 nM and 2400 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.