Learn More
TARGETMOL CHEMICALS INC RACLOPRIDE 25MG
SDP
Catalog No. 502576302
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Raclopride is a selective antagonist of dopamine D2/D3 receptor. purity: 98%
Catalog No. 50-257-6302 Supplier TARGETMOL CHEMICALS INC Supplier No. T763525MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Raclopride is a selective antagonist of dopamine D2/D3 receptor. purity: 98%