Learn More
Medchemexpress LLC HY-103414 5mg Medchemexpress, Raclopride CAS:84225-95-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1034145MG
Encompass_Preferred
Medchemexpress, HY-103414 5mg Raclopride CAS:84225-95-6 Raclopride is a dopamine D 2 / D 3 receptor antagonist, which binds to D 2 and D 3 receptor s with dissociation constants ( K i s) of 1.8 nM and 3.5 nM, respectively, but has a very low affinity for D 1 and D 4 receptors with K i s of 18000 nM and 2400 nM, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2244