Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Raclopride-d5 hydrochloride | 1217623-85-2 | 99.8% | 352.27 | C15H16D5Cl3N2O3 | 1 MG
SDP

Catalog No. 5000246552
Encompass_Preferred

Raclopride-d5 hydrochloride is the deuterium-labeled (d5) isotopologue of raclopride supplied as the hydrochloride salt and intended for use as an analytical reference standard or stable-isotope internal standard for quantitative assays.

  • Deuterated raclopride (d5) for use as an internal standard.
  • Hydrochloride salt in solid form for stable handling.
  • High purity (99.82%) suitable for analytical applications.
  • Molecular weight 352.27 and formula C15H16D5Cl3N2O3.
  • Available in small reference sizes (1 mg) for trace-level assays.
  • Certificate of analysis and safety data sheet available for quality verification.

Catalog No. 50-002-46552 Supplier Medchemexpress LLC Supplier No. HY103414S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Raclopride-d5 hydrochloride is the deuterium-labeled (d5) isotopologue of raclopride supplied as the hydrochloride salt and intended for use as an analytical reference standard or stable-isotope internal standard for quantitative assays.

  • Deuterated raclopride (d5) for use as an internal standard.
  • Hydrochloride salt in solid form for stable handling.
  • High purity (99.82%) suitable for analytical applications.
  • Molecular weight 352.27 and formula C15H16D5Cl3N2O3.
  • Available in small reference sizes (1 mg) for trace-level assays.
  • Certificate of analysis and safety data sheet available for quality verification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.