Learn More
Cayman Chemical Racanisodamine, Purity: ≥98%, CAS: 17659-49-3, 100 mg
SDP
Catalog No. 502982434
Encompass_Preferred
A weak antagonist of α1-adrenoceptors, blocking WB-4101 and clonidine binding in brain membrane preparations with pKi values of 2.63 and 1.61, respectively; also has antioxidant effects
Catalog No. 50-298-2434 Supplier Cayman Chemical Supplier No. 19650100 MG
A weak antagonist of α1-adrenoceptors, blocking WB-4101 and clonidine binding in brain membrane preparations with pKi values of 2.63 and 1.61, respectively; also has antioxidant effects