Learn More
Medchemexpress LLC Ractopamine hydrochloride | 90274-24-1 | 99.2% | 50 MG
SDP
Catalog No. 5000413404
Encompass_Preferred
Ractopamine hydrochloride, also known as LY031537, is a potent β-adrenergic receptor (βAR) agonist with a Kd value of approximately 25 nM for pig β1AR and β2AR. It is associated with protein metabolism and is used in research to promote lean tissue growth and enhance production efficiency in pigs.
  • Potent β-adrenergic receptor (βAR) agonist
  • Impacts protein metabolism
  • Research application in lean tissue growth
  • Improved production efficiency in pigs
Catalog No. 50-004-13404 Supplier Medchemexpress LLC Supplier No. HYB142150MG
Ractopamine hydrochloride, also known as LY031537, is a potent β-adrenergic receptor (βAR) agonist with a Kd value of approximately 25 nM for pig β1AR and β2AR. It is associated with protein metabolism and is used in research to promote lean tissue growth and enhance production efficiency in pigs.
  • Potent β-adrenergic receptor (βAR) agonist
  • Impacts protein metabolism
  • Research application in lean tissue growth
  • Improved production efficiency in pigs