Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ractopamine ketone-d6 (hydrochloride) | 1185012-24-1 | 95.2% | 341.86 | 1 MG
SDP

Catalog No. 5000347935
Encompass_Preferred

This product is the deuterium-labeled form of 1-(4-Hydroxyphenyl)-2-((4-(4-hydroxyphenyl)butan-2-yl)amino)ethanone hydrochloride. It is a stable, solid chemical intended for laboratory use.

  • Deuterium-labeled form of 1-(4-hydroxyphenyl)-2-((4-(4-hydroxyphenyl)butan-2-yl)amino)ethanone hydrochloride
  • Stable under recommended storage conditions
  • Appearance: White to off-white solid
  • Purity (HPLC): 95.2%
  • Isotopic enrichment: 98.9%
  • Recommended storage: Powder -20°C for 3 years; in solvent -80°C for 6 months, -20°C for 1 month
  • Not classified as a hazardous substance or mixture
  • Not dangerous goods for transport

Catalog No. 50-003-47935 Supplier Medchemexpress LLC Supplier No. HY139687S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is the deuterium-labeled form of 1-(4-Hydroxyphenyl)-2-((4-(4-hydroxyphenyl)butan-2-yl)amino)ethanone hydrochloride. It is a stable, solid chemical intended for laboratory use.

  • Deuterium-labeled form of 1-(4-hydroxyphenyl)-2-((4-(4-hydroxyphenyl)butan-2-yl)amino)ethanone hydrochloride
  • Stable under recommended storage conditions
  • Appearance: White to off-white solid
  • Purity (HPLC): 95.2%
  • Isotopic enrichment: 98.9%
  • Recommended storage: Powder -20°C for 3 years; in solvent -80°C for 6 months, -20°C for 1 month
  • Not classified as a hazardous substance or mixture
  • Not dangerous goods for transport

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.