Learn More
Research Products International Corp Polyethylene Bags, Unprinted, Clear, 4 mil Thick, 10 x 12 Inches, 100 per Case
SDP
Catalog No. 50213085
Encompass
Heat Sealable Polyethylene Bags are transparent and specially designed for use in all hybridization applications. These bags are especially useful as specimen bags or for product storage. Bags are heavy duty and are 4 mil thick. Bags perform well at cryogenic temperatures (-80°C) and can be flash frozen in liquid nitrogen -196°C. Ideal for quickly sealing solutions with a bag sealer that is also available from RPI.
Catalog No. 50-213-085 Supplier Research Products International Corp Supplier No. 286669C
Heat Sealable Polyethylene Bags are transparent and specially designed for use in all hybridization applications. These bags are especially useful as specimen bags or for product storage. Bags are heavy duty and are 4 mil thick. Bags perform well at cryogenic temperatures (-80°C) and can be flash frozen in liquid nitrogen -196°C. Ideal for quickly sealing solutions with a bag sealer that is also available from RPI.