Learn More
Research Products International Corp Radioactive Waste Disposal Bags, 3 Mil Thick, 24 x 36 Inches, Yellow Tint, Pre-Printed with Caution Message, 250 per Roll
SDP
Catalog No. 50213094
Encompass
Radioactive material waste bags are available in a variety of convenient sizes and styles to help sort and identify different types of waste. Strong, bottom-sealed waste liners are free of heavy metals, making them safe for landfill or incineration.
Catalog No. 50-213-094 Supplier Research Products International Corp Supplier No. 365107Y
Radioactive material waste bags are available in a variety of convenient sizes and styles to help sort and identify different types of waste. Strong, bottom-sealed waste liners are free of heavy metals, making them safe for landfill or incineration.