Learn More
Research Products International Corp AC Adapter and Charger, 120/18V, For use with Radiation and Contamination Monitor, Models GM1, GM2 and SD-10
SDP
Catalog No. 50213054
Encompass
Model SD10 is supplied with a high sensitivity scintillation probe for detecting low energy gamma emitters such as Iodine-125. The detector utilizes a high quality photomultiplier tube coupled to a 1'' x 2 mm thick sodium iodide crystal with a thin 1 mil (7 mg/cm2) aluminum window. Also suitable for thyroid monitoring of 125I uptake.
Optional power supplies are also available.
Catalog No. 50-213-054 Supplier Research Products International Corp Supplier No. 950112
Model SD10 is supplied with a high sensitivity scintillation probe for detecting low energy gamma emitters such as Iodine-125. The detector utilizes a high quality photomultiplier tube coupled to a 1'' x 2 mm thick sodium iodide crystal with a thin 1 mil (7 mg/cm2) aluminum window. Also suitable for thyroid monitoring of 125I uptake.
Optional power supplies are also available.