Learn More
Aviva Systems Biology Corp RAD1 Antibody - middle region (ARP57717_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57717_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAD1. It was validated on Western Blot. RAD1,RAD1 homolog (S. pombe),REC1; HRAD1; RAD1,cell cycle checkpoint protein RAD1,cell cycle checkpoint protein RAD1,OTTHUMP00000115992,exonuclease homolog RAD1,DNA repair exonuclease REC1,checkpoint control protein HRAD1,DNA repair exonuclease rad1 homolog,cell cycle checkpoint protein Hrad1,rad1-like DNA damage checkpoint proteinThe immunogen is a synthetic peptide directed towards the middle region of human RAD1. Alternate names: HRAD1, REC1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-360