Learn More
Medchemexpress LLC Rad16-I hydrochloride | 2100275-49-6 | 98.9% | 10 MG
SDP
Catalog No. 5000424401
Encompass_Preferred
RAD16-I hydrochloride is a soft nanofibrous self-assembling peptide. It provides a suitable microenvironment for the proliferation and differentiation of human mesenchymal stem cells (hMSC) into chondrocytes. It is also an ionic complementary peptide that has been used as a model to investigate the potential amyloid-like staining properties of Self-Assembling Peptide Nanofibers (SAPNFs).
  • Provides a suitable microenvironment for human mesenchymal stem cells' (hMSC) proliferation and differentiation into chondrocytes.
  • Used to check potential amyloid-like staining properties of SAPNFs.
Catalog No. 50-004-24401 Supplier Medchemexpress LLC Supplier No. HYP2632A10MG
RAD16-I hydrochloride is a soft nanofibrous self-assembling peptide. It provides a suitable microenvironment for the proliferation and differentiation of human mesenchymal stem cells (hMSC) into chondrocytes. It is also an ionic complementary peptide that has been used as a model to investigate the potential amyloid-like staining properties of Self-Assembling Peptide Nanofibers (SAPNFs).
  • Provides a suitable microenvironment for human mesenchymal stem cells' (hMSC) proliferation and differentiation into chondrocytes.
  • Used to check potential amyloid-like staining properties of SAPNFs.