Learn More
Medchemexpress LLC Rad16-I (hydrochloride) | 2100275-49-6 | 98.9% | 5 MG
SDP
Catalog No. 5000424151
Encompass_Preferred
RAD16-I hydrochloride is a soft nanofibrous self-assembling peptide. It provides a suitable microenvironment for human mesenchymal stem cells' (hMSC) proliferation and differentiation into chondrocytes. This ionic complementary peptide is also used as a model to check potential amyloid-like staining properties of self-assembling peptide nanofibers (SAPNFs).
  • Provides a suitable microenvironment for human mesenchymal stem cells' (hMSC) proliferation and differentiation into chondrocytes.
  • Used as a model to check potential amyloid-like staining properties of self-assembling peptide nanofibers (SAPNFs).
  • Intended for research use only.
  • Suitable for laboratory use.
Catalog No. 50-004-24151 Supplier Medchemexpress LLC Supplier No. HYP2632A5MG
RAD16-I hydrochloride is a soft nanofibrous self-assembling peptide. It provides a suitable microenvironment for human mesenchymal stem cells' (hMSC) proliferation and differentiation into chondrocytes. This ionic complementary peptide is also used as a model to check potential amyloid-like staining properties of self-assembling peptide nanofibers (SAPNFs).
  • Provides a suitable microenvironment for human mesenchymal stem cells' (hMSC) proliferation and differentiation into chondrocytes.
  • Used as a model to check potential amyloid-like staining properties of self-assembling peptide nanofibers (SAPNFs).
  • Intended for research use only.
  • Suitable for laboratory use.