Learn More
Aviva Systems Biology Corp RAD17 Antibody - C-terminal region (ARP30206_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30206_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RAD17. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RAD17. Alternate names: CCYC, HRAD17, R24L, RAD17Sp, Rad24, RAD24, RAD17SPApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-54142