Learn More
Aviva Systems Biology Corp RAD17 Antibody - N-terminal region (ARP30546_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30546_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAD17. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human RAD17. Alternate names: CCYC, FLJ41520, HRAD17, R24L, RAD17SP, RAD24ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-53879