Learn More
Aviva Systems Biology Corp RAD23A Antibody - middle region (ARP42211_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42211_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RAD23A. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human RAD23A. Alternate names: HHR23A, MGC111083ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58145