Learn More
Aviva Systems Biology Corp RAD23B Antibody - middle region (ARP56506_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56506_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAD23B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RAD23B. Alternate names: HHR23B, HR23B, P58ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61885