Learn More
Research Products International Corp RAD-5 Lock Box, 1/2 Inch Thick, 11 x 7 x 5 Inches Outside Dimensions
SDP
Catalog No. 50550437
Encompass
The Original Locking Container for Radioactive Materials
Rad-Lock Box containers provide secure storage for radioactive materials in the laboratory. Made from thick, heavy-duty acrylic, each unit features a built-in cylinder key lock and tamper-resistant hinges to restrict access to authorized personnel. Designed for convenience, they safely store, handle, and transport small bottles, vials, and tubes. The spacious interior can be packed with crushed ice to keep samples cold and is freezer-safe to –70 °C. Rad-Lock Boxes attenuate beta emissions from isotopes such as P-32, C-14, H-3, and S-35, helping protect users from beta radiation.
Catalog No. 50-550-437 Supplier Research Products International Corp Supplier No. RLB15
The Original Locking Container for Radioactive Materials
Rad-Lock Box containers provide secure storage for radioactive materials in the laboratory. Made from thick, heavy-duty acrylic, each unit features a built-in cylinder key lock and tamper-resistant hinges to restrict access to authorized personnel. Designed for convenience, they safely store, handle, and transport small bottles, vials, and tubes. The spacious interior can be packed with crushed ice to keep samples cold and is freezer-safe to –70 °C. Rad-Lock Boxes attenuate beta emissions from isotopes such as P-32, C-14, H-3, and S-35, helping protect users from beta radiation.