Learn More
Aviva Systems Biology Corp RAD51 Antibody - N-terminal region (ARP33450_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33450_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAD51. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RAD51. Alternate names: BRCC5, HRAD51, HsRad51, HsT16930, RAD51A, RECAApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-55031
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.