Learn More
Medchemexpress LLC Rad51-in-1 | 2101739-18-6 | 99.9% | 50 MG
SDP
Catalog No. 5000231355
Encompass_Preferred
RAD51-IN-1 is a small-molecule inhibitor of the RAD51 recombinase, derived from B02, intended for preclinical cancer research to inhibit homologous recombination and reduce RAD51 foci formation in tumor cells.
  • Inhibits RAD51-mediated homologous recombination.
  • Displays cytotoxic activity in multiple human cancer cell lines with low micromolar IC50 values.
  • Reported high purity (99.89%) and solid, yellow to orange physical form.
  • Stable as a powder under recommended storage (powder: -20°C for years).
  • Available in small solid quantities and as a 10 mM solution in DMSO for in vitro use.
Catalog No. 50-002-31355 Supplier Medchemexpress LLC Supplier No. HY12270550MG
RAD51-IN-1 is a small-molecule inhibitor of the RAD51 recombinase, derived from B02, intended for preclinical cancer research to inhibit homologous recombination and reduce RAD51 foci formation in tumor cells.
  • Inhibits RAD51-mediated homologous recombination.
  • Displays cytotoxic activity in multiple human cancer cell lines with low micromolar IC50 values.
  • Reported high purity (99.89%) and solid, yellow to orange physical form.
  • Stable as a powder under recommended storage (powder: -20°C for years).
  • Available in small solid quantities and as a 10 mM solution in DMSO for in vitro use.