Learn More
Medchemexpress LLC HY-101462 10mg , RAD51 Inhibitor B02 CAS:1290541-46-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10146210MG
Encompass_Preferred
HY-101462 10mg RAD51 Inhibitor B02 CAS: 1290541-46-6 RAD51 Inhibitor B02 (B02) is an inhibitor of human RAD51 with an IC50 of 27.4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9885