Learn More
Apexbio Technology LLC RAD51 Inhibitor B02, 10mg CAS# 1290541-46-6
SDP
Supplier:  Apexbio Technology LLC B839610
Encompass_Preferred
RAD51 inhibitor B02 (B02) inhibits human RAD51 with IC50 of 27.4 M. In vitro, the RAD51 inhibitor B02 specifically inhibits human RAD51 (IC50 = 27.4 M), but does not inhibit its E. coli homologous RecA (IC50> 250 M). In vivo, B02 can significantly enhance the therapeutic effect of cisplatin on tumor cells. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8446