Learn More
Genetex Inc Rad51 antibody [N1C2], 100 uL
SDP
Supplier:  Genetex Inc GTX100469
Encompass
RAD51 antibody detects RAD51 protein, also known as RAD51 homolog A. RAD51 (37 kDa) is the indispensable strand exchange factor for homologous recombination in DNA repair. It is also essential for replication fork processing and for genome stability. This RAD51 antibody is a well-cited rabbit polyclonal validated for many applications.
Catalog No. NC1814566