Learn More
Aviva Systems Biology Corp RAD51AP1 Antibody - middle region (ARP40653_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40653_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAD51AP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RAD51AP1. Alternate names: PIR51ApplicationWB. Species reactivity: Dog, Horse, Human, Rabbit, Rat
Catalog No. 50-284-885