Learn More
Aviva Systems Biology Corp RAD51L1 Antibody - middle region (ARP57721_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57721_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAD51L1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RAD51L1. Alternate names: MGC34245, R51H2, RAD51B, REC2, hREC2, RAD51L1ApplicationWB. Species reactivity: Cow, Human, Mouse
Catalog No. 50-286-362