Learn More
Aviva Systems Biology Corp RAD54B Antibody - N-terminal region (ARP36406_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36406_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAD54B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RAD54B. Alternate names: FSBP, RDH54ApplicationWB. Species reactivity: Cow, Human, Rabbit, Rat
Catalog No. 50-014-56141