Learn More
Aviva Systems Biology Corp RAD54L Antibody - N-terminal region (ARP36355_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36355_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RAD54L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RAD54L. Alternate names: HR54, RAD54A, hHR54, hRAD54ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56125