Learn More
Sino Biological Rad6 / UBE2A Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 12726T08100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Rad6 / UBE2A (rh Rad6 / UBE2A; Catalog#12726-H07E; P49459; Met1-Cys152). Rad6 / UBE2A specific IgG was purified by Human Rad6 / UBE2A affinity chromatography.
Catalog No. 50-224-7962