Learn More
Medchemexpress LLC Radafaxine hydrochloride | 106083-71-0 | 99.01% | 292.20 | 1 ML
SDP
Catalog No. 5000387522
Encompass_Preferred
Radafaxine hydrochloride (GW-353162A) is an inhibitor of DAT (dopamine transporter) and NET (norepinephrine transporter) and a modulator of the nAChR family. It is a potent metabolite of bupropion and selectively inhibits the reuptake of norepinephrine over dopamine.
  • Inhibits DAT (dopamine transporter) and NET (norepinephrine transporter).
  • Modulates the nAChR family.
  • Potent metabolite of bupropion.
  • Selective for inhibiting norepinephrine reuptake over dopamine.
  • Low potency in blocking DAT and slow blockade suggest unlikelihood of reinforcing effects.
Catalog No. 50-003-87522 Supplier Medchemexpress LLC Supplier No. HY1759010MM/1ML
Add to Cart
Add to Cart
Radafaxine hydrochloride (GW-353162A) is an inhibitor of DAT (dopamine transporter) and NET (norepinephrine transporter) and a modulator of the nAChR family. It is a potent metabolite of bupropion and selectively inhibits the reuptake of norepinephrine over dopamine.
  • Inhibits DAT (dopamine transporter) and NET (norepinephrine transporter).
  • Modulates the nAChR family.
  • Potent metabolite of bupropion.
  • Selective for inhibiting norepinephrine reuptake over dopamine.
  • Low potency in blocking DAT and slow blockade suggest unlikelihood of reinforcing effects.