Learn More
Thermo Scientific™ RadEye SPRD with Large Display
GSA_VA
Catalog No. 17000848 Shop All Thermo Scientific Products
Change view
Click to view available options
Energy Range:
58 keV to 3 MeV
Catalog No. Energy Range
17-000-848 58 keV to 3 MeV
1 options
This item is not returnable. View return policy
Catalog No. 17-000-848 Supplier Thermo Scientific™ Supplier No. 4250827
Add to Cart
Add to Cart
This item is not returnable. View return policy
Pocket-sized Personal Radiation Detector includes CSI gamma detector with integral SPM.
Full nuclide identification with indication and accurate energy compensated.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Energy Range 58 keV to 3 MeV
Product Type Personal Radiation Detector
Measurement Range 1 μR/h to 25 mR/h