Learn More
Apexbio Technology LLC Radezolid, 10mg. Cas: 869884-78-6 MFCD: MFCD22628873. Novel oxazolidinone antibiotic agent
SDP
Supplier:  Apexbio Technology LLC A374910
Encompass_Preferred
Radezolid Description:MIC90: Radezolid was approximately four-times more potent than linezolid against MRSA, with MIC90 of 0.5 mg/l and 2.0 mg/l, respectively [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4833