Learn More
Thermo Scientific™ RadHalo RDP Portable Spectroscopic Area Monitors
Catalog No. 19175808
Change view
Click to view available options
Includes:
Neutron, Radio
Neutron, Satellite, Radio
Radio
Satellite, Radio
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-175-808 Neutron, Satellite, Radio
19-175-809 Neutron, Radio
19-175-810 Satellite, Radio
19-175-811 Radio
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-175-808 Supplier Thermo Scientific™ Supplier No. RH RDPNSR
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Identify radiation quickly and accurately, in any environment
The Thermo Scientific RadHalo RDP PortableSpectroscopic Area Monitor is designed for portability, rapid deployment, and autonomous operation. It is ideal for monitoring special events and radiological emergencies. Long 60 hour battery can be charged via AC mains or solar panel. Multiple wireless transmission methods insure instrument communications in nearly any scenario. Monitor and control unit remotely from PC or mobile device.
  • Rapid dose rate response
  • Immediate accurate identification
  • Rugged design
  • Autonomous operation anywhere
  • Secure transmissions
  • Detector Fusion Capable
TRUSTED_SUSTAINABILITY
Specifications
Detector Type Spectroscopic Detector: 3 x 3 in. NaI
Product Type Spectroscopic Area Monitor
Description RadHalo RDP-NS
Communications Wi-Fi, 2.4 GHz Long Range Radio, Cellular Reachback
Includes Neutron, Satellite, Radio