Learn More
Thermo Scientific™ RadHalo Spectroscopic Area Monitor Antenna Kits
GSA_VA
Catalog No. 19175829 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
2.4 GHz Directional Antenna Kit
2.4 GHz Omnidirectional Antenna Kit
GSM Cellular External Antenna Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-175-829 2.4 GHz Omnidirectional Antenna Kit
19-175-827 GSM Cellular External Antenna Kit
19-175-828 2.4 GHz Directional Antenna Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-175-829 Supplier Thermo Scientific™ Supplier No. COMA001628
Add to Cart
Add to Cart
For use with RadHalo FM Spectroscopic Area Monitors
GSM Cellular External Antenna Kit - Includes Includes antenna for GSM Cellular and 10 ft. connection cable to RadHalo FM. 2.4 GHz Directional Antenna Kit - Includes high gain directional flat antenna and 10 ft. connection cable to RadHalo FM or Matrix Base Radio and Repeater. 2.4 GHz Omnidirectional Antenna Kit - Includes high gain omnidirectional pole style antenna and 10 ft. connection cable to RadHalo FM or Matrix Radio and Repeater.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Product Type 2.4 GHz Omnidirectional Antenna Kit