Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ RadHalo Spectroscopic Area Monitor Radio Communication Options
GSA_VA

Catalog No. 19175826 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Matrix Base Radio - Single Laird 2.4GHz radio in IP67 rated housing
Matrix Base Radio and Matrix Radio Repeater Pole Mount Kit
Matrix Radio Repeater - Dual 2.4 GHz Laird Radios
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-175-826 Matrix Base Radio and Matrix Radio Repeater Pole Mount Kit
19-175-824 Matrix Base Radio - Single Laird 2.4GHz radio in IP67 rated housing
19-175-825 Matrix Radio Repeater - Dual 2.4 GHz Laird Radios
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-175-826 Supplier Thermo Scientific™ Supplier No. COMC004162
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with RadHalo Spectroscopic Area Monitor models with the radio option

Matrix Base Radio and Matrix Radio Repeater
  • Include rugged IP67 housing, 2.4 GHz radio, external antenna connection, power connection, ethernet connection and wall mount
  • Require an appropriate 2.4 GHz antenna kit
  • Do not include a pole mounting kit or screws/anchors for wall mount
The Matrix Base Radio and Matrix Radio Repeater Pole Mount Kit includes complete kit for mounting radios to a pole up to 12 inches in diameter.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Matrix Base Radio and Matrix Radio Repeater Pole Mount Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.