Learn More
Stoelting™ Radial 8-Arm Maze
SDP
Catalog No. 10000303 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Mouse
Catalog No. Animal
10-000-303 Mouse
1 options
Catalog No. 10-000-303 Supplier Stoelting™ Supplier No. 60150
Add to Cart
Add to Cart
Stoelting™ Radial 8-Arm Maze is for use in evaluating spatial learning and memory.
Apparatus consists of eight equally spaced arms radiating from a small circular central platform. The design ensures that after checking for food at the end of an arm the animal is forced to return to the central platform before making another choice. As a result, the animal always has eight possible options.
Specifications
For Use With (Application) Neuroscience
Animal Mouse