Learn More
Thermo Scientific Ceramic D-Torch Kits for the iCAP Series ICP-OES Systems | | 1 EA
Catalog No. NC1971460 Shop All Thermo Scientific Products
Encompass
Reduce the cost of consumables traditionally associated with ICP-OES analysis using these demountable Thermo Scientific Ceramic D-Torch Kits for Thermo Scientific iCAP Series ICP-OES systems in place of a conventional quartz torch. These torch kits help prevent potential devitrification and significantly extend torch lifespan.
Catalog No. NC1971460 Supplier Thermo Scientific Supplier No. 842312052201
Reduce the cost of consumables traditionally associated with ICP-OES analysis using these demountable Thermo Scientific Ceramic D-Torch Kits for Thermo Scientific iCAP Series ICP-OES systems in place of a conventional quartz torch. These torch kits help prevent potential devitrification and significantly extend torch lifespan.