Learn More
Perkin Elmer US LLC Radial Purge Extension Quartz Window for Avio 500
Catalog No. 502104725
Encompass_Preferred
Replaceable quartz window that slides into the purge extension between the torch and the optics.
Catalog No. 50-210-4725 Supplier Perkin Elmer US LLC Supplier No. N0810346
Replaceable quartz window that slides into the purge extension between the torch and the optics.