Learn More
Perkin Elmer US LLC Radial Purge Extension Window for Optima 4x00/5x00 DV
Catalog No. 509043778
Encompass_Preferred
The Radial View Purge Extension Window is a replaceable quartz window that slides into the mount between the torch and the optics for the radial view.
Catalog No. 50-904-3778 Supplier Perkin Elmer US LLC Supplier No. N0770322
The Radial View Purge Extension Window is a replaceable quartz window that slides into the mount between the torch and the optics for the radial view.