Learn More
Waters Corp RADIAN ASAP Direct Mass Detector, Single Quadrupole, Integrated ASAP Source
Catalog No. 502391400 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The RADIAN ASAP Direct Mass Detector is a dedicated direct analysis system for rapid, easy, and low-cost liquid and solid analysis. It uses single quadrupole mass spectrometry with an Atmospheric Pressure Solids Analysis Probe (ASAP) for quality and ease of use in a compact footprint. This enables direct MS analysis without complex sample preparation or MS experience.
  • Simple operation and data interpretation
  • Minimal sample preparation; no separation required
  • Real-time results and classification
  • Easy to learn, fully automated setup
  • Direct sampling onto glass probe; minimizes solvent use
  • Rapid 30-second analysis time
  • Compact design
  • Dedicated single quadrupole mass detector with integrated ASAP source
  • Pre-optimized ASAP source for structural identification
  • Automated mass calibration and resolution verification
  • Programmable temperature control
Catalog No. 50-239-1400 Supplier Waters Corp Supplier No. 201000330
The RADIAN ASAP Direct Mass Detector is a dedicated direct analysis system for rapid, easy, and low-cost liquid and solid analysis. It uses single quadrupole mass spectrometry with an Atmospheric Pressure Solids Analysis Probe (ASAP) for quality and ease of use in a compact footprint. This enables direct MS analysis without complex sample preparation or MS experience.
  • Simple operation and data interpretation
  • Minimal sample preparation; no separation required
  • Real-time results and classification
  • Easy to learn, fully automated setup
  • Direct sampling onto glass probe; minimizes solvent use
  • Rapid 30-second analysis time
  • Compact design
  • Dedicated single quadrupole mass detector with integrated ASAP source
  • Pre-optimized ASAP source for structural identification
  • Automated mass calibration and resolution verification
  • Programmable temperature control