Learn More
MSC Radians All-Purpose Synthetic Leather Glove, Medium, Reinforced Palm, 1 Pair
Catalog No. 5000802341
Encompass
This all-purpose synthetic leather glove offers superior toughness and comfort. It features a reinforced padded palm, reinforced fingertips, and double stitching for enhanced durability in various applications.
  • Terry cloth backhand for wiping sweat away from brow
  • Breathable neoprene between fingers offers improved flexibility
  • Neoprene knuckle strap provides extra dexterity
  • Reinforced fingertips for longer wear
  • Reinforced thumb saddle provides greater protection and toughness
  • Streamlined elastic cuff with hook and loop closure ensures a secure fit
  • Neoprene guard in the wrist system for extra protection from debris
  • Breathable spandex backhand stretches for maximum comfort, fit, and ventilation
  • Double stitching for greater durability
  • Concealed interior seam stitching prevents snagging
Catalog No. 50-008-02341 Supplier MSC Supplier No. 01163559
This all-purpose synthetic leather glove offers superior toughness and comfort. It features a reinforced padded palm, reinforced fingertips, and double stitching for enhanced durability in various applications.
  • Terry cloth backhand for wiping sweat away from brow
  • Breathable neoprene between fingers offers improved flexibility
  • Neoprene knuckle strap provides extra dexterity
  • Reinforced fingertips for longer wear
  • Reinforced thumb saddle provides greater protection and toughness
  • Streamlined elastic cuff with hook and loop closure ensures a secure fit
  • Neoprene guard in the wrist system for extra protection from debris
  • Breathable spandex backhand stretches for maximum comfort, fit, and ventilation
  • Double stitching for greater durability
  • Concealed interior seam stitching prevents snagging