Learn More
MSC Radians AXIS Cut Protection Glove, A4, Foam Nitrile, 1 Pair
Catalog No. 5000766300
Encompass
The Radians AXIS™ A4 Foam Nitrile Coated Cut Protection Glove offers superior protection against abrasion, cuts, tear, and puncture. Its seamless 13 gauge HPPE shell with fiberglass and foam nitrile dipped palm ensures comfort and protection in various applications.
  • Provides cut resistance with 13 gauge HPPE.
  • Foam nitrile palm dip protects from abrasion in wet or dry conditions.
  • Seamless design ensures a comfortable fit.
  • Meets EN 388:2003 standards (4 5 4 4).
  • ANSI Cut Level A4, EN388 Cut Level 5.
Catalog No. 50-007-66300 Supplier MSC Supplier No. 90122888
The Radians AXIS™ A4 Foam Nitrile Coated Cut Protection Glove offers superior protection against abrasion, cuts, tear, and puncture. Its seamless 13 gauge HPPE shell with fiberglass and foam nitrile dipped palm ensures comfort and protection in various applications.
  • Provides cut resistance with 13 gauge HPPE.
  • Foam nitrile palm dip protects from abrasion in wet or dry conditions.
  • Seamless design ensures a comfortable fit.
  • Meets EN 388:2003 standards (4 5 4 4).
  • ANSI Cut Level A4, EN388 Cut Level 5.