Learn More
Grainger Radians Chemical Splash Safety Goggle, Anti-Fog, Indirect Vent, 24/Pk.
Catalog No. 502786765
Encompass_Preferred
These chemical splash safety goggles are designed to provide reliable eye protection in laboratory and industrial environments. Featuring a clear frame and lens, these goggles are engineered to withstand chemical hazards while maintaining high optical clarity.
  • Indirect vent design provides protection against chemical splashes
  • Anti-fog lens coating ensures clear vision in demanding conditions
  • Fits over most prescription eyewear for complete versatility
  • Clear frame and lens construction for optimal visibility
Catalog No. 50-278-6765 Supplier Grainger Supplier No. 199JP5
These chemical splash safety goggles are designed to provide reliable eye protection in laboratory and industrial environments. Featuring a clear frame and lens, these goggles are engineered to withstand chemical hazards while maintaining high optical clarity.
  • Indirect vent design provides protection against chemical splashes
  • Anti-fog lens coating ensures clear vision in demanding conditions
  • Fits over most prescription eyewear for complete versatility
  • Clear frame and lens construction for optimal visibility