Learn More
MSC Radians Crossfire Infinity Safety Glasses, ANSI Z87.1, 1 ea
Catalog No. 5000687571
Encompass
Crossfire Infinity Safety Glasses feature a full frame design, offering a comfortable fit suitable for all-day wear. These safety glasses are available with various frame and lens options, including polarized lenses. Their tough polycarbonate lenses provide protection against 99.9% of harmful UV rays and meet ANSI Z87.1-2015 standards.
  • Vented rubber nosepiece for enhanced breathability
  • Dual mold, rubber temple grips for improved stability
  • Option for polarized lenses
  • Hard coat polycarbonate lens resists scratches
  • Offers 99.9% UV protection
  • Meets ANSI Z87.1 standards
Catalog No. 50-006-87571 Supplier MSC Supplier No. 53893772
Crossfire Infinity Safety Glasses feature a full frame design, offering a comfortable fit suitable for all-day wear. These safety glasses are available with various frame and lens options, including polarized lenses. Their tough polycarbonate lenses provide protection against 99.9% of harmful UV rays and meet ANSI Z87.1-2015 standards.
  • Vented rubber nosepiece for enhanced breathability
  • Dual mold, rubber temple grips for improved stability
  • Option for polarized lenses
  • Hard coat polycarbonate lens resists scratches
  • Offers 99.9% UV protection
  • Meets ANSI Z87.1 standards