Learn More
MSC Radians Cut Glove, ANSI A2 Cut, Polyurethane Palm, 1 ea
Catalog No. 5000769463
Encompass
The Radians RWG550, part of the Ghost™ Series, is a cut protection glove designed for comfort in hazardous applications. It features a 13-gauge Trosar by DSM™ shell, with fibers manufactured in the USA, and a polyurethane palm coating. This glove is suitable for work environments with sharp objects and is often utilized in sectors like HVAC, Metal Fabrication, and Manufacturing.
  • Features a 13-gauge Trosar by DSM™ shell with USA manufactured fibers
  • Polyurethane palm dip for abrasion protection in both wet and dry conditions
  • Seamless design for a comfortable fit
  • Tested to EN 388 standards (4 X 4 2 B) for abrasion, blade cut, tear, and puncture
  • ANSI rated cut level A2, abrasion 6, puncture 3
  • Suitable for HVAC, metal fabrication, and manufacturing environments with sharp objects
Catalog No. 50-007-69463 Supplier MSC Supplier No. 59281295
The Radians RWG550, part of the Ghost™ Series, is a cut protection glove designed for comfort in hazardous applications. It features a 13-gauge Trosar by DSM™ shell, with fibers manufactured in the USA, and a polyurethane palm coating. This glove is suitable for work environments with sharp objects and is often utilized in sectors like HVAC, Metal Fabrication, and Manufacturing.
  • Features a 13-gauge Trosar by DSM™ shell with USA manufactured fibers
  • Polyurethane palm dip for abrasion protection in both wet and dry conditions
  • Seamless design for a comfortable fit
  • Tested to EN 388 standards (4 X 4 2 B) for abrasion, blade cut, tear, and puncture
  • ANSI rated cut level A2, abrasion 6, puncture 3
  • Suitable for HVAC, metal fabrication, and manufacturing environments with sharp objects