Learn More
MSC Radians Cut Protection Glove, HPPE, A2 Cut, 1 Pair
Catalog No. 5000794588
Encompass
The Radians Radwear Silver Series is a high-visibility gripper glove designed for cut protection. It features an HPPE high-visibility shell that offers durability and blade cut resistance, an elastic cuff for a comfortable fit, and a clear, water-based PUD coated palm to ensure a slip-free grip in various applications.
  • ANSI Cut Level A2 for reliable protection
  • High-visibility HPPE shell for enhanced safety
  • Clear water-based PUD coated palm provides secure grip
  • Elastic slip-on cuff for comfort and ease of wear
  • Compliant with FDA 21 CFR 177.152 for food processing applications
  • Meets EN 388:2003 standards (4 3 4 4) for abrasion, cut, tear, and puncture resistance
  • Seamless lining and washable design
Catalog No. 50-007-94588 Supplier MSC Supplier No. 69555696
The Radians Radwear Silver Series is a high-visibility gripper glove designed for cut protection. It features an HPPE high-visibility shell that offers durability and blade cut resistance, an elastic cuff for a comfortable fit, and a clear, water-based PUD coated palm to ensure a slip-free grip in various applications.
  • ANSI Cut Level A2 for reliable protection
  • High-visibility HPPE shell for enhanced safety
  • Clear water-based PUD coated palm provides secure grip
  • Elastic slip-on cuff for comfort and ease of wear
  • Compliant with FDA 21 CFR 177.152 for food processing applications
  • Meets EN 388:2003 standards (4 3 4 4) for abrasion, cut, tear, and puncture resistance
  • Seamless lining and washable design