Learn More
Grainger Radians Face Shield, Polycarbonate, Clear, 8 in. x 15.5 in., 1 ea
Catalog No. 502786496
Encompass_Preferred
This clear polycarbonate face shield provides essential eye and face protection for a variety of work environments. Designed for durability and clarity, the shield meets rigorous industry safety standards while offering universal compatibility with most hard hats and faceshield systems.
  • Constructed from durable 0.04 in. thick clear polycarbonate
  • Features universal pegs to accommodate most hard hat systems
  • Meets ANSI Z87.1+ safety standards
  • Provides 99.9% ultraviolet ray protection (UVA, UVB, and UVC)
  • Measures 8 in. x 15.5 in.
Catalog No. 50-278-6496 Supplier Grainger Supplier No. 199TY8
This clear polycarbonate face shield provides essential eye and face protection for a variety of work environments. Designed for durability and clarity, the shield meets rigorous industry safety standards while offering universal compatibility with most hard hats and faceshield systems.
  • Constructed from durable 0.04 in. thick clear polycarbonate
  • Features universal pegs to accommodate most hard hat systems
  • Meets ANSI Z87.1+ safety standards
  • Provides 99.9% ultraviolet ray protection (UVA, UVB, and UVC)
  • Measures 8 in. x 15.5 in.