Learn More
Grainger Radians Galaxy Safety Eyewear, Polycarbonate, Clear, ANSI Z87.1+, 12/Pk.
Catalog No. 502786883
Encompass_Preferred
This item has to be ordered in multiples of 12 per supplier requirements.
These Galaxy™ safety glasses feature a lightweight, one-piece polycarbonate lens design with integral side shields and an extended brow guard for comprehensive orbital protection. Engineered for comfort and versatility, these glasses include 4-position adjustable temple arms to provide a secure, universal fit for various users. The impact-resistant lenses provide 99.9% protection against harmful UVA and UVB radiation, making them suitable for diverse indoor work environments.
  • Meets ANSI Z87.1+ impact standards
  • Hard-coated scratch-resistant lens surface
  • Integrated side shields for enhanced lateral protection
  • Lightweight wraparound frame style
  • Includes 4-position adjustable length temples
  • Provides 99.9% UVA/UVB protection
Catalog No. 50-278-6883 Supplier Grainger Supplier No. 199JT0
This item has to be ordered in multiples of 12 per supplier requirements.
These Galaxy™ safety glasses feature a lightweight, one-piece polycarbonate lens design with integral side shields and an extended brow guard for comprehensive orbital protection. Engineered for comfort and versatility, these glasses include 4-position adjustable temple arms to provide a secure, universal fit for various users. The impact-resistant lenses provide 99.9% protection against harmful UVA and UVB radiation, making them suitable for diverse indoor work environments.
  • Meets ANSI Z87.1+ impact standards
  • Hard-coated scratch-resistant lens surface
  • Integrated side shields for enhanced lateral protection
  • Lightweight wraparound frame style
  • Includes 4-position adjustable length temples
  • Provides 99.9% UVA/UVB protection