Learn More
MSC Radians High Visibility Vest, 3X-Large, Hook and Loop, 1 ea
Catalog No. 5000793903
Encompass
This high visibility vest is designed for safety and convenience in various work environments. It features a durable construction and multiple safety elements to ensure worker visibility.
  • Three X-large size
  • Green color
  • Hook and loop closure
  • Three pockets
  • Mesh and solid material composition
  • One horizontal silver stripe, 2 in wide
  • ISEA 107 and ANSI/ISEA 107 approved
  • Five point breakaway design
Catalog No. 50-007-93903 Supplier MSC Supplier No. 69580298
This high visibility vest is designed for safety and convenience in various work environments. It features a durable construction and multiple safety elements to ensure worker visibility.
  • Three X-large size
  • Green color
  • Hook and loop closure
  • Three pockets
  • Mesh and solid material composition
  • One horizontal silver stripe, 2 in wide
  • ISEA 107 and ANSI/ISEA 107 approved
  • Five point breakaway design