Learn More
Grainger Radians Journey Ballistic Safety Glasses, IRUV 5.0, Universal, 12/pk
Catalog No. 502786759
Encompass_Preferred
This item has to be ordered in multiples of 12 per supplier requirements.
These Journey® safety glasses are designed for high-performance optical clarity and reliable protective coverage. Featuring a half-frame, wraparound style, these glasses provide 99.9% UVA/UVB protection and are ballistic tested for enhanced durability. The polycarbonate lenses are hard-coated for superior scratch resistance, ensuring long-lasting visibility in demanding work environments.
  • 4-position telescoping temples for superior fit
  • 9.75 base lens provides excellent optics and coverage
  • Rubber nose piece allows for all day comfort
  • ANSI Z87.1+ certified for impact resistance
  • Hard coated lenses for scratch resistance
  • Ballistic tested for high-impact protection
Catalog No. 50-278-6759 Supplier Grainger Supplier No. 199HY7
This item has to be ordered in multiples of 12 per supplier requirements.
These Journey® safety glasses are designed for high-performance optical clarity and reliable protective coverage. Featuring a half-frame, wraparound style, these glasses provide 99.9% UVA/UVB protection and are ballistic tested for enhanced durability. The polycarbonate lenses are hard-coated for superior scratch resistance, ensuring long-lasting visibility in demanding work environments.
  • 4-position telescoping temples for superior fit
  • 9.75 base lens provides excellent optics and coverage
  • Rubber nose piece allows for all day comfort
  • ANSI Z87.1+ certified for impact resistance
  • Hard coated lenses for scratch resistance
  • Ballistic tested for high-impact protection