Learn More
Grainger Radians Mirage Bifocal Safety Glasses, Polycarbonate, Clear, +1.5 Diopter, 1/Each
Catalog No. 502785879
Encompass_Preferred
This item has to be ordered in multiples of 12 per supplier requirements.
These bifocal safety glasses provide eye protection with integrated magnification, featuring a clear, distortion-free polycarbonate lens designed to assist with close-up tasks. The sleek, lightweight construction ensures a comfortable fit for diverse users, while the vented rubber nosepiece enhances breathability during extended wear.
  • Bifocal lens for clear close-up vision
  • Hard coat polycarbonate lens protects against scratches
  • Distortion-free optically correct lens
  • Vented rubber nosepiece for comfort and breathability
  • Complies with ANSI/ISEA Z87.1-2020 standards
  • Impact-rated to Z87+
  • Provides 99.9% UVA and UVB protection
Catalog No. 50-278-5879 Supplier Grainger Supplier No. 199XE0
This item has to be ordered in multiples of 12 per supplier requirements.
These bifocal safety glasses provide eye protection with integrated magnification, featuring a clear, distortion-free polycarbonate lens designed to assist with close-up tasks. The sleek, lightweight construction ensures a comfortable fit for diverse users, while the vented rubber nosepiece enhances breathability during extended wear.
  • Bifocal lens for clear close-up vision
  • Hard coat polycarbonate lens protects against scratches
  • Distortion-free optically correct lens
  • Vented rubber nosepiece for comfort and breathability
  • Complies with ANSI/ISEA Z87.1-2020 standards
  • Impact-rated to Z87+
  • Provides 99.9% UVA and UVB protection